Learn More
PSI-6130, MedChemExpress
Product Code. 16456207
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
2mg
50mg
5mg
Product Code. 16456207
Brand: Medchem Express HY10165
MedChemExpress PSI-6130 is a potent and selective inhibitor of HCV NS5B polymerase, and inhibits HCV replication with a mean IC50 of 0.6 μM.
PSI-6130 is a potent and selective inhibitor of HCV NS5B polymerase, and inhibits HCV replication with a mean IC50 of 0.6 μM.
Specifications
Chemical Name or Material PSI-6130
CAS 817204-33-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C10H14FN3O4
Quantity 5 mg
Synonym R 1656
Solubility Information DMSO : 25 mg/mL (96.44 mM; Need ultrasonic)
SMILES O=C1N=C(C=CN1[C@@H]([C@@](C)([C@@H]2O)F)O[C@@H]2CO)N
Molecular Weight (g/mol) 259.23
Formula Weight 259.23
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less
Research purposes only