missing translation for 'onlineSavingsMsg'
Learn More

PSI-6130, MedChemExpress

Product Code. 16403488
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16403488

Brand: Medchem Express HY10165

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PSI-6130 is a potent and selective inhibitor of HCV NS5B polymerase, and inhibits HCV replication with a mean IC50 of 0.6 μM.

PSI-6130 is a potent and selective inhibitor of HCV NS5B polymerase, and inhibits HCV replication with a mean IC50 of 0.6 μM.

Specifications

Chemical Name or Material PSI-6130
CAS 817204-33-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C10H14FN3O4
Quantity 10 mg
Synonym R 1656
Solubility Information DMSO : 25 mg/mL (96.44 mM; Need ultrasonic)
SMILES O=C1N=C(C=CN1[C@@H]([C@@](C)([C@@H]2O)F)O[C@@H]2CO)N
Molecular Weight (g/mol) 259.23
Formula Weight 259.23
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.