Learn More
PRUNE2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11458-1-AP-20UL
 View more versions of this product
Product Code. 16828553
  • € 145.00 / 20µL
Description
The calculated molecular weight of this protein is 340 kDa. Our antibody specifically recognizes the 140-150 kDa band which is an isoform or cleaved fragment of PRUNE2.
PRUNE2 gene, also named BMCC1, encodes protein prune homolog 2. PRUNE2 gene are overlapping with PCA3, one of the most prostate cancer specific markers. PRUNE2 may play an important role in regulating differentiation, survival and aggressiveness of the tumor cells. Besides its high level of expression seen in the nervous system (brain, cerebellum and spinal cord), it is also expressed at high levels in noneuroblastoma, rhabdomyosarcoma, melanoma and some osteosarcoma cell lines, whereas at only low levels in cancer cell lines of liver, breast, thyroid and colon. The Calculated molecular weight of this protein is 340 kDa.
Specifications
Specifications
PRUNE2
Polyclonal
Unconjugated
PRUNE2
A214N16.3, BMCC1, BNIPXL, C9orf65, KIAA0367, Protein prune homolog 2, prune homolog 2 (Drosophila), PRUNE2, RP11 58J3.2
Rabbit
Antigen Affinity Chromatography
RUO
158471, 293823, 353211
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.26 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q52KR3, Q5BJR4, Q8WUY3
Prune2
PRUNE2 Fusion Protein Ag2025
20 μL
Primary
Rat, Mouse, Human