Learn More
Hamilton™ PRP-1, 30-50 μm Bulk Resin
PRP-1 columns have inherent reversed-phase separation qualities that eliminate the requirement for a stationary phase coating. Hydrolysis of the stationary phase coating is often the limiting factor in column lifetime, so the lack of coating on the PRP-1 resin extends the longevity of the column. PRP-1 also have exceptional sample recovery (≥ 95%).
Brand: Hamilton™ 79582
Description
Specification Sheet
- Separation Mode – Reversed-Phase
- Stationary Phase – PRP-1
- Format – Bulk Resin
- Particle Size – 30-50 μm
- Packing Material Type – PSDVB
- Pore Size – 100 Å
- Max Pressure – 5,000 psi
- Restoration – Run a gradient of 100% water to 100% acetonitrile (Repeat three times)
- UOM – per gram
Specifications
30 to 50 μm |