Learn More
Christ Martin™ Protective Net
Brand:  Christ Martin™ 127382
Product Code. 15220226
  • € 12.31 / Each
Explore more special offers
Description
Description
The version with LDplus control is particularly suited to standard applications in round-bottomed or similar flasks and small quantities in product trays and other containers. For advanced applications requiring more process control and/or temperature-controlled shelves for the product, we recommend the LSCplus range. Almost all models are available with ice condenser temperatures of approx. -55 °C (main application: aqueous solutions) and -85 °C (for products with low freezing points, e.g. solvents).