missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terracycle™ Protective Eyewear Zero Waste Box™

Click to view available options
Size:
Small
Medium
Large
Unit Size:
Each
Description
Use this Zero Waste Box to recycle:
- Swimming goggles, tanning and safety glasses
Approximate waste units accepted:
- Small: 200 units
- Medium: 510 units
- Large: 1000 units
Do not send in:
- Sunscreen bottles
- Other protective equipment
- Medical waste
- This includes material contaminated with blood or bodily fluids and waste that originate from health care facilities, hospitals, home care, physicians' offices, dental offices, blood banks, veterinary clinics, research laboratories, etc.
- Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic) which presents a danger to the environment, or to people
- Batteries, pressurized canisters, broken glass and medical waste
Specifications
Specifications
| Capacity (Metric) | 15 kg |
| Capacity (English) | 33 lb. |
| Color | Beige, Green, Red, White |
| Dimensions (L x W x H) | 30.5 x 30.5 x 84 cm |
| Disposable | Single-use |
| Latex Free | Yes |
| Size | Medium |
| Sterility | Non-sterile |
| Autoclavable | Not Autoclavable |
| Material | Cardboard, Plastic |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction