Learn More
Mutexil™ Protective Overshoe Coverings
Product Code. 12340180
Click to view available options
Product Type:
Overshoe
Protective CPE Visitor Overshoes
Sticky Overshoe
Color:
Blue
Transparent
White
Size:
2X-Large
Universal
X-Large
Unit Size:
25 pairs
Pack of 1000
Pack of 2000
Pack of 300
Product Code. 12340180
Brand: Mutexil™ 4579911
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Choose from a range of choices and construction, from sticky, anti-slip, fluid -resistant, polypropylene and CPE variations with Mutexil™ Protective Overshoe Coverings.
  • Various sizes and colors
  • Needs-based styles
  • Per new standards EN ISO 20345 and EN ISO 20347
TRUSTED_SUSTAINABILITY
Specifications
Color Transparent
Length (Metric) 400 mm
Material Tyvek
Seam Type Welded
Size Universal
Sole Features Antislip
Width (Metric) 450 mm
Disposable Single-use
Packaging Quantity 1000/Pk.
Sterility Sterile
Thickness 70μm
Resistant To Fluid
Product Type Overshoe