Learn More
Mutexil™ Protective Overshoe Coverings
Product Code. 11780114
Click to view available options
Product Type:
Overshoe
Protective CPE Visitor Overshoes
Sticky Overshoe
Color:
Blue
Transparent
White
Size:
2X-Large
Universal
X-Large
Unit Size:
25 pairs
Pack of 1000
Pack of 2000
Pack of 300
Product Code. 11780114
Brand: Mutexil™ 4579911
Only null left
Add to basket
Add to basket
Choose from a range of choices and construction, from sticky, anti-slip, fluid -resistant, polypropylene and CPE variations with Mutexil™ Protective Overshoe Coverings.
  • Various sizes and colors
  • Needs-based styles
  • Per new standards EN ISO 20345 and EN ISO 20347
Specifications
Color Transparent
Length (Metric) 400 mm
Material Tyvek
Seam Type Welded
Size Universal
Sole Features Antislip
Width (Metric) 450 mm
Disposable Single-use
Packaging Quantity 1000/Pk.
Sterility Sterile
Thickness 70μm
Resistant To Fluid
Product Type Overshoe