Learn More
Thermo Scientific™ ProSpecT™ Adenovirus Microplate Assay
Rapidly detect Adenovirus specific antigen from human fecal samples.
Brand: Thermo Scientific™ R240096
Description
Rapidly detect Adenovirus specific antigen in fresh or preserved human fecal specimens using Thermo Scientific™ ProSpecT™ Adenovirus Microplate Assay.
Manage your resources, increase productivity and improve workflow efficiency with this direct, qualitative enzyme immmunoassay for the detection of Adenovirus. It combines excellent sensitivity/specificity, a common procedure with no indeterminate range, plus flexible specimen transport options.- Excellent clinical performance
- Cary Blair transport option which saves a dilution step
- Results within 100 minutes
- Room temperature incubation
- True colorless negatives
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
2°C to 8°C | |
ProSpecT Adenovirus Microplate Assay | |
Transfer pipettes, microplate strip holder and cover, microplate (8 wells/strip), enzyme conjugate, positive control, negative control, sample diluent, wash buffer, color substrate, stop solution and procedure card | |
96 Tests/Kit |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
3443996 | Certificate of Analysis | 14/05/2022 | R240096 |
3416592 | Certificate of Analysis | 31/03/2022 | R240096 |
3326423 | Certificate of Analysis | 27/01/2022 | R240096 |
3282611 | Certificate of Analysis | 01/07/2021 | R240096 |
3240410 | Certificate of Analysis | 27/04/2021 | R240096 |