Learn More
Invitrogen™ ProQuantum™ Software v1.1
Description
ProQuantum Software enables you to analyze ProQuantum high-sensitivity immunoassay results acquired from any qPCR instrument. The analysis features include standard curve qualification with built-in outlier detection, interpolation of unknown samples for quantitation, and group-wise statistical analysis. Step-by-step guidance is provided for preparation of an assay run, including setting up the standard curve and plate layout for 96- or 384-well plates.
• Convenient setup wizard to minimize errors―automatic calculations for standard curve serial dilution setup
• Built-in statistical comparison―easily compare controls vs treatment groups
• Export capabilities―view results in Excel and PDF, or conveniently formatted for GraphPad Prism
ProQuantum Software consists of two modules: Create Experiment and Import and View Results.
• Create Experiment allows you to set up a standard curve, create a plate layout, and assign descriptions and groups to individual samples wells
• Import and View Results allows you to import experimental data adjusted to fit the standard curve on a 4PL or 5PL weighted regression algorithm, interpolate unknown protein concentrations, view heat maps, perform group-wise comparisons, and export the data
ProQuantum Software is compatible with all ProQuantum immunoassay kits and with all Applied Biosystems qPCR instruments, including QuantStudio 3/5/6 Flex/7 Flex/12K Flex, StepOne, StepOnePlus, 7500, 7500 Fast, ViiA 7, or other compatible qPCR instruments.
The preferred operating system is Windows 7 or 10.
For more information, visit thermofisher.com/proquantum.
Specifications
Specifications
| Type | ProQuantum Software v1.0 |
| For Use With (Equipment) | 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, BioRad CFX96™, QuantStudio™, ViiA™ 7 System, StepOnePlus™ System, Roche LightCycler 480, Bio-Rad CFX384 Touch™, Bio-Rad CFX Connect™, StepOne™ System, Bio-Rad CFX96 Touch™ |
| Product Line | ProQuantum |
| Quantity | 5 License Keys |
| Content And Storage | Includes 5 license keys for 5 users |
| Software | Data Analysis Software |