Learn More
Propylene carbonate, 99.5%, Thermo Scientific Chemicals
Propylene carbonate, 99.5%, Acros Organics, Quantity: 2.5L, Packaging: Glass bottle, Boiling Point: 240.0 deg.C to 243.0 deg.C, Undesignated, Melting Point: -49.0 deg.C, Molecular Weight: 102.09, Percent Purity: 99.5%, Assay Percent Range: 99.4% min. (GC), CAS: 108-32-7, Density: 1.2040g/mL
€ 40.10 - € 345.72
Chemical Identifiers
CAS | 108-32-7 |
---|---|
Molecular Formula | C4H6O3 |
Molecular Weight (g/mol) | 102.09 |
MDL Number | MFCD00005385,MFCD00798264,MFCD00798265 |
InChI Key | RUOJZAUFBMNUDX-UHFFFAOYNA-N |
Synonym | propylene carbonate, 1,2-propylene carbonate, 1,2-propanediol cyclic carbonate, texacar pc, 1,3-dioxolan-2-one, 4-methyl, cyclic propylene carbonate, arconate 5000, 1,2-propanediol carbonate, 1-methylethylene carbonate, cyclic 1,2-propylene carbonate |
PubChem CID | 7924 |
IUPAC Name | 4-methyl-1,3-dioxolan-2-one |
SMILES | CC1COC(=O)O1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10625572
|
Thermo Scientific Acros
131565000 |
500 mL | Glass bottle |
€ 40.10
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10046612
|
Thermo Scientific Acros
131560010 |
1 L | Glass bottle |
€ 62.21
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10615572
|
Thermo Scientific Acros
131560025 |
2.5 L | Glass bottle |
€ 122.61
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10669263
|
Thermo Scientific Acros
131560100 |
10 L | Plastic drum |
€ 345.72
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
108-32-7 | |
102.09 | |
RUOJZAUFBMNUDX-UHFFFAOYNA-N | |
7924 | |
CC1COC(=O)O1 |
C4H6O3 | |
MFCD00005385,MFCD00798264,MFCD00798265 | |
propylene carbonate, 1,2-propylene carbonate, 1,2-propanediol cyclic carbonate, texacar pc, 1,3-dioxolan-2-one, 4-methyl, cyclic propylene carbonate, arconate 5000, 1,2-propanediol carbonate, 1-methylethylene carbonate, cyclic 1,2-propylene carbonate | |
4-methyl-1,3-dioxolan-2-one |
Specifications
108-32-7 | |
1.2040g/mL | |
116°C | |
99.4% min. (GC) | |
1.4200 to 1.4220 | |
1.204 | |
Solubility in water: 200g/L (25°C). Other solubilities: miscible with acetone,benzene,chloroform,dmf,,ether,ethyl acetate and ethyl alcohol,slightly soluble in carbon tetrachloride,soluble in most organic solvents | |
CC1COC(=O)O1 | |
102.09 | |
2.8 mPa.s (20°C) | |
99.5% | |
Propylene carbonate |
-49.0°C | |
240.0°C to 243.0°C | |
Authentic | |
C4H6O3 | |
MFCD00005385,MFCD00798264,MFCD00798265 | |
propylene carbonate, 1,2-propylene carbonate, 1,2-propanediol cyclic carbonate, texacar pc, 1,3-dioxolan-2-one, 4-methyl, cyclic propylene carbonate, arconate 5000, 1,2-propanediol carbonate, 1-methylethylene carbonate, cyclic 1,2-propylene carbonate | |
RUOJZAUFBMNUDX-UHFFFAOYNA-N | |
4-methyl-1,3-dioxolan-2-one | |
7924 | |
102.09 | |
Liquid |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 203-572-1
- Propylene carbonate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only