missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress Prolylleucine
MedChemExpress Prolylleucine is a dipeptide containing branched-chain amino acids. Prolylleucine can affect the circadian rhythms and behaviour of animals.
Brand: MedChemExpress HY-112173
This item is not returnable.
View return policy
Specifications
99.82% | |
Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month | |
362.42 g/mol | |
25 mg | |
DMSO : ≥ 100 mg/mL (275.92 mM) |
61596-47-2 | |
Neuroscience-Neuromodulation | |
Peptides | |
Room temperature | |
((Benzyloxy)carbonyl)-L-prolyl-D-leucine |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction