missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ VersaTouch™ 92-465 Series Blue Nitrile Splash Gloves

Product Code. 10472794 Shop All Ansell Products
Click to view available options
Size:
7
7.5/8
8
8.5/9
9.5/10
Unit Size:
100 pairs
Pack of 100
Pack of 1000
This item is not returnable. View return policy

Product Code. 10472794

Brand: Ansell™ 92465_8.59

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Provides outstanding resistance to animal fats compared with PVC or natural rubber. Ansell VersaTouchTM 92-465 Series Blue Nitrile Splash Gloves are manufactured from an innovative nitrile compound which offers excellent chemical and mechanical properties, comfort and dexterity.

  • Single use splash resistant gloves
  • Textured finish on fingertips. Improved grip
  • Contains no natural rubber
  • Suitable for contact with all fatty foods
  • Good resistance to animal fats compared with PVC or natural rubber
  • Powder-free
  • No risk of contaminating food with corn starch used to powder gloves

Applications:

  • All final stages of meat or fish processing
  • Controlled-environment rooms
  • Prepared meals
  • Dairy processing
  • Catering
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 420:2003 + I1:2009 and EN 374:2003
Accelerator Free No
Applications Industrial
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish Fingertip Textured
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Nitrile Gloves
Size 8.5/9
Sterility Non-sterile
Thickness (English) 4.7 mil
Thickness (Metric) 0.12 mm
Glove Cuff Style Beaded/Rolled
Model 924659
Series 92-465
Length (Metric) 240 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.