Learn More
Ansell™ VersaTouch™ 92-465 Series Blue Nitrile Splash Gloves
Product Code. 10152214 Shop All Ansell Products
Click to view available options
Size:
7
7.5/8
8
8.5/9
9.5/10
Unit Size:
100 pairs
Pack of 100
Pack of 1000
Product Code. 10152214
Brand: Ansell™ 92465_6.57
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Provides outstanding resistance to animal fats compared with PVC or natural rubber. Ansell VersaTouchTM 92-465 Series Blue Nitrile Splash Gloves are manufactured from an innovative nitrile compound which offers excellent chemical and mechanical properties, comfort and dexterity.
  • Contains no natural rubber
  • Suitable for contact with all fatty foods
  • Textured finish on fingertips for an improved grip
  • Powder-free - no risk of contaminating food with corn startch used to powder gloves
  • Ideal for all final stages of meat and fish processing, controlled environment rooms, dairy processing and catering
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 420:2003 + I1:2009 and EN 374:2003
Accelerator Free No
Applications Industrial
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish Fingertip Textured
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Nitrile Gloves
Size 7
Sterility Non-sterile
Thickness (English) 4.7 mil
Thickness (Metric) 0.12 mm
Model 924657
Series 92-465