Learn More
Invitrogen™ ProcartaPlex Porcine Basic Kit
Ideal solution for assessing multiple protein biomarkers in a single sample. This kit includes all non-target specific reagents - assay diluent, wash buffers, SA-PE and plates. Explore our broad portfolio of pre-configures panels and combinable single analytes.
Brand:  Invitrogen™ EPX010-60460-901
276.45 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Product Code. 15542137
  • € 291.00 / 96 tests
Explore more special offers
Includes:
  • Reading Buffer
  • Wash Buffer 10x
  • Streptavidin-PE
  • Procine Specific Universal Assay Buffer
  • Detection Antibody Diluent
  • PCR 8-Tube Strip
  • Adhesive Film
  • Greiner Microplate black 96 well
Description
Description
ProcartaPlex™ immunoassays are our ideal solution for assessing multiple protein biomarkers in a single sample.
Our ProcartaPlex assays use Luminex ™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins.
ProcartaPlex™ assays are subject to the same stringent and robust development and manufacturing as are our extensively validated Platinum and Instant ELISA kits. Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma.
Explore our broad portfolio of pre-configures panels and combinable single analytes.
For combination of only Simplex Kits an additional Basic Kit is required. This kit includes all non-target specific reagents - assay diluent, wash buffers, SA-PE and plates.
This kit does not contain a Multiscreen Filter Plate.
Reported Application
Multiplex Immunoassay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Basic and Setup Kits
Porcine
See Certificate of Analysis
ProcartaPlex™
96 Tests
Combinable
Luminex™ Instruments