Learn More
Invitrogen™ ProcartaPlex Porcine Basic Kit
Ideal solution for assessing multiple protein biomarkers in a single sample. This kit includes all non-target specific reagents - assay diluent, wash buffers, SA-PE and plates. Explore our broad portfolio of pre-configures panels and combinable single analytes.
Brand: Invitrogen™ EPX010-60460-901
276.45 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Includes:
- Reading Buffer
- Wash Buffer 10x
- Streptavidin-PE
- Procine Specific Universal Assay Buffer
- Detection Antibody Diluent
- PCR 8-Tube Strip
- Adhesive Film
- Greiner Microplate black 96 well
Description
ProcartaPlex™ immunoassays are our ideal solution for assessing multiple protein biomarkers in a single sample.
Our ProcartaPlex assays use Luminex ™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins.
ProcartaPlex™ assays are subject to the same stringent and robust development and manufacturing as are our extensively validated Platinum and Instant ELISA kits. Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma.
Explore our broad portfolio of pre-configures panels and combinable single analytes.
For combination of only Simplex Kits an additional Basic Kit is required. This kit includes all non-target specific reagents - assay diluent, wash buffers, SA-PE and plates.
This kit does not contain a Multiscreen Filter Plate.
Reported Application
Multiplex Immunoassay
Specifications
Basic and Setup Kits | |
Porcine | |
See Certificate of Analysis | |
ProcartaPlex™ |
96 Tests | |
Combinable | |
Luminex™ Instruments |
For Research Use Only. Not for use in diagnostic procedures.