Learn More
Invitrogen™ ProcartaPlex Mouse Basic Kit
Product Code. 15502137
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
Product Code. 15502137
Brand: Invitrogen™ EPX01020440901
Includes
  • Reading Buffer
  • Wash Buffer 10x
  • Streptavidin-PE
  • Universal Assay Buffer
  • Detection Antibody Diluent Mouse
  • PCR 8-Tube Strip
  • Adhesive Film
  • Greiner Microplate black 96 well
Ideal solution for assessing multiple protein biomarkers in a single sample. This kit includes all non-target specific reagents - assay diluent, wash buffers, SA-PE and plates. Explore our broad portfolio of pre-configures panels and combinable single analytes.
  • Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
  • This kit does not contain a Multiscreen Filter Plate
  • ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
TRUSTED_SUSTAINABILITY
Specifications
Product Type Basic and Setup Kits
Quantity 96 Tests
Species Mouse
Combinability Combinable
Assay Sensitivity See Certificate of Analysis
For Use With (Equipment) Luminex™ Instruments
Product Line ProcartaPlex