missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Pritelivir
GREENER_CHOICE

Product Code. 18715666
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 18715666

Brand: Tocris Bioscience™ 7321/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective herpes simplex virus (HSV) helicase primase inhibitor

Pritelivir is a potent and selective herpes simplex virus (HSV) helicase primase inhibitor. Pritelivir inhibits viral replication in Vero cells (IC50 = 0.02 μM for HSV-1 and HSV-2). It also reduces β-amyloid and P-tau levels in Vero cells. Pritelivir is active against clinical isolates and porcine and bovine HSV strains but displays reduced activity against varicella zoster virus and cytomegalovirus in vitro. In addition, the compound exhibits antiviral activity in HSV infected mice.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[5-(Aminosulfonyl)-4-methyl-2-thiazolyl]-N-methyl-4-(2-pyridinyl)benzeneacetamide
CAS 348086-71-5
Quantity 50 mg
Target Antiviral Compounds
Molecular Formula C18H18N4O3S2
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.