Learn More
Epredia™ PrintMate™ AS Cassette Printer
The small footprint of the Epredia™ SlideMateTM AS Slide Printer brings the convenience of on-demand slide printing directly to the microtome. The SlideMate AS Slide Printer drastically reduces the chance for patient identification errors by allowing the laboratory to print accurate information at the moment the slide is required.
€ 30110.00 - € 70910.00
Product Code | Brand | Description | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Includes | Price | Quantity & Availability | |||||
15488891
|
Epredia™
A83800001 |
PrintMate AS 150 Cassette Printer | PrintMate AS USB cable Power cable 2 cassette hoppers One roll of tape (installed) Epredia™ LabWriter™ software CD Operator guide |
€ 30110.00
Each |
In Stock
Log in to see stock. |
|||||
15498891
|
Epredia™
A83800002 |
PrintMate AS 450 Cassette Printer | PrintMate AS USB cable Power cable 6 cassette hoppers One roll of tape (installed) Epredia™ LabWriter™ software CD Operator guide |
€ 40835.00
Each |
In Stock
Log in to see stock. |
|||||
15408901
|
Epredia™
A83800003 |
PrintMate AS 900 Cassette Printer | PrintMate AS USB cable Power cable 12 cassette hoppers One roll of tape (installed) Epredia™ LabWriter™ software CD Operator guide |
€ 65290.00
Each |
Estimated Shipment: 07-04-2025
Log in to see stock. |
|||||
15418901
|
Epredia™
A83800004 |
PrintMate AS 150 Cassette Printing System | PrintMate AS USB cable Power cable 2 cassette hoppers One roll of tape (installed) Epredia™ LabWriter™ software CD Operator guide All-in-one PC Digital imager |
€ 31845.00
Each |
Estimated Shipment: 07-04-2025
Log in to see stock. |
|||||
15441021
|
Epredia™
A83800005 |
PrintMate AS 450 Cassette Printing System | PrintMate AS USB cable Power cable 6 cassette hoppers One roll of tape (installed) Epredia™ LabWriter™ software CD Operator guide All-in-one PC Digital imager |
€ 43810.00
Each |
Estimated Shipment: 07-04-2025
Log in to see stock. |
|||||
15448901
|
Epredia™
A83800006 |
PrintMate AS 900 Cassette Printing System | PrintMate AS USB cable Power cable 12 cassette hoppers One roll of tape (installed) Epredia™ LabWriter™ software CD Operator guide All-in-one PC Digital imager |
€ 70910.00
Each |
Estimated Shipment: 07-04-2025
Log in to see stock. |
|||||
Description
Highlights:- Prints each cassette in 10 seconds or less
- Small footprint allows the printer to fit easily next to the grossing station
- Print only the cassettes you need, right at the point of use
- Print barcodes on cassettes to automate downstream processes
- Eliminate manual handwriting processes
Specifications
100 to 240 VAC, 50/60 Hz, 150 VA (512 BTU/h) |
Product Title
Epredia™ PrintMate™ AS Cassette Printer