Learn More
Invitrogen™ PRIMER S-OLIGO 200 NMOL
Custom Primer
Brand:  Invitrogen™ 10917037
5.00 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Product Code. 10674763
  • € 6.66 / Each
Explore more special offers
Description
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
S-Oligo Primer, 200 nmol
S-Oligo Primer
200 nmol