Learn More
Invitrogen™ PRIMER INOSINE 25 NMOL
Custom Primer
Brand: Invitrogen™ 10624062
7.18 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).Specifications
Normal or Reversed-phase Chromatography, HPLC or PAGE | |
Custom Inosine Primer | |
HPLC |
Custom Inosine Primer, 25 nmole, Purified by Normal or Reversed-phase Chromatography, HPLC or PAGE, Used to Design a Custom DNA Oligonucleotide, With any of a Variety of 3' and 5' Modification | |
25 nmol | |
Design a Custom DNA Oligonucleotide, with any of a variety of 3' and 5' Modification |