Learn More
JULABO PRESTO Process Systems
Click to view available options
Model:
A38
A41
A70
W41
Unit Size:
Each
Description
- Wide working temperature ranges without changing fluids
- Ethernet interface
- SD-Card slot
- pace optimized design creates more room directly next to the units
- Connections for USB, Ethernet, RS232, and Alarm output
Specifications
Specifications
| Temperature Range (Metric) | -75°C to +250°C |
| Dimensions (L x W x H) | 29.3 x 22.4 x 34.6 in. |
| Model | A70 |
| Type | Process System |