Learn More
Ohaus™ Precision High Capacity Balance
Product Code. 15183959 Shop All Ohaus Products
Click to view available options
Capacity:
12 kg
24 kg
35 kg
Model:
EX12001M-EU
EX24001M-EU
EX35001M-EU
Unit Size:
Each
Product Code. 15183959
Brand: Ohaus™ 30057069
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Simplify even the most complex high-capacity laboratory measurements with this balance. Ohaus™ Precision High Capacity Balances provide unmatched functionality in a line of high-performance, high-capacity balances.
  • Display: 5.7 inches (145 mm) Full-color VGA graphic touchscreen display with user-controlled brightness
  • AC Adapter (included) or rechargeable battery (accessory sold separately)
  • RS232 and USB (included) or second RS232/Ethernet (accessories sold separately) . GLP/GMP data output with real-time clock.
  • Modular design, IP54 metal base, Die-cast top housing, stainless steel pan, up-front level indicator, integral weigh below hook, calibration lock, two touchless sensors, full housing in-use cover
  • Selectable environmental filters, auto tare, user selectable span calibration points, software lockout and reset menu, user selectable communication settings and data print options, user definable project and user IDs, auto standby
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance OIML F1 or ASTM Class 2
Connection Type Standard RS232, 2x USB, Optional 2nd RS232 or Ethe
Capacity 35 kg
Display Type 5.7 in. Full-color VGA Graphic Touchscreen
Readability 0.1 g
For Use With (Application) In laboratory, in concrete industry, in assembly-l
IP Rating IP54
Leveling Thumbwheels need to be adjusted to level the balan
Linearity ±0.2 g
Material Die-cast Metal Bottom Housing
Model EX35001M-EU
Repeatability ±0.1 g
Temperature Range (Metric) 10°C to 30°C
Type High Capacity Precision Balance
Weighing Modes Weighing, Parts Counting, Percent Weighing, Check
Dimensions (D x W x H) 443 x 377 × 120 mm
Width (Metric) 377 mm
Width (Metric) Pan 377 mm
Height (Metric) 120 mm
Depth (Metric) 443 mm
Depth (Metric) Pan 311 mm
Weight (Metric) 10 kg
Electrical Requirements Power Input 100 to 240VAC, 0.5 to 0.25A, 47 to 63H
Voltage 100/240 V
Wattage 15 w
Amperage 0.63 A
Frequency 47/63 Hz