Learn More
OHAUS™ Precision Balance - PR Series
Product Code. 16670324 Shop All Ohaus Products
Click to view available options
Model:
PR1602M
PR2202M
PR223M
PR4201M
PR4202M
PR423M
PR523M
PR6201M
Unit Size:
Each
Product Code. 16670324
Brand: OHAUS™ 30524387
Designed for Routine Weighing Applications in Your Workplace
  • The PR is equipped with 3 essential weighing modes, RS-232 connectivity for data transfer and storage, and internal calibration (in select models), making it ideal for routine weighing applications.
  • Equipped with an easy-to-read, bright backlit display and a simple user interface, the PR is incredibly easy to operate, with almost no training required.
  • The PR's small footprint saves desktop space while providing a large weighing surface. The PR is durably constructed, and features a stainless steel pan to withstand day-to-day use in the workplace.
TRUSTED_SUSTAINABILITY
Specifications
Connection Type RS232 (one port included)
Description User-selectable environmental filters and brightness settings, auto-tare, auto-dim, user-selectable span calibration points, software lockout and reset menu, user-selectable communication settings and data print options, user-definable project and user IDs, software overload/underload indicator, stability indicator
Capacity 420.00 g
Display Type Backlit LCD
Readability 0.0010 g
For Use With (Application) Basic Weighing, Parts Counting, Percent Weighing
Linearity 0.0020 g
Model PR423M
Repeatability 0.0010 g
Stabilization Time 2.000 s
Weighing Modes Milligram, Gram, & Carat
Dimensions (L x W x H) 51.000 x 39.000 x 53.000 cm
Length (English) 12.50 in.
Length (Metric) 317.00 mm
Width (English) 7.90 in.
Width (Metric) 201.00 mm
Height (English) 11.90 in.
Height (Metric) 303.000 mm
Depth (English) 12.50 in.
Depth (Metric) 317.00 mm
Weight (English) 10.000 lb.
Weight (Metric) 4.500 kg
Electrical Requirements AC Adapter (included)