missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Pre-cut Vinyl Labels for BradyJet J4000 and J5000

Product Code. 17419192
Click to view available options
Height (Metric):
55.1 mm
74.9 mm
104.9 mm
177.8 mm
210 mm
Vertical Repeat (Metric):
92.1 mm
117.5 mm
162 mm
222.3 mm
266.7 mm
Width (Metric):
85.1 mm
110 mm
155 mm
254 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17419192

Brand: Brady™ 148659

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

B-2569 is ideal for indoor facility labels, including 5S, safety, warning, pipe markers, equipment and visual workplace labels on stainless steel and smooth surfaces.

  • Specially-formulated topcoat enhances colors and label appearance
  • Tested to withstand exposure to liquids, alcohol and chemicals, as well as indoor fluorescent and UV lighting
  • Topcoat protects print from smudging and harsh conditions
  • Inkjet-receptive labels for use with the BradyJet J4000 or J5000 Color Label Printer
  • An ink cartridge is required to print this material

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance Compliant with Directive 2011/65/EU and 2015/863
Adhesive Type Acrylic, Permanent
Coating None
Color White
Finish Matte
Format Die-cut
Material Vinyl (Inkjet-receptive)
Printable Sides Single-side Printable
Print Technology Ink Jet
Product Type Label
Sterility Non-sterile
For Use With (Application) Electrical Labeling, Hazard Communication Identification, Warehouse Marking
For Use With (Equipment) BradyJet J4000 and J5000 Color Label Printers
Labels per Row 1
Min. Operating Temperature (Metric) -40°C
No. per Pack 100 Labels/Roll
Quantity 100 Labels
Shape Rectangle
Temperature Range (English) -40°F to +140°F
Temperature Range (Metric) -40°C to +60°C
Thickness (English) 0.0053 in.
Thickness (Metric) 0.14 mm
Vertical Repeat (Metric) 266.7 mm
Dimensions (LxW) 254 x 177.80 mm
Length (English) 10 in.
Width (English) 7 in.
Width (Metric) 254 mm
Width (Metric) Printable Area 254 mm
Height (English) 10 in.
Height (Metric) 177.8 mm
Height (Metric) Printable Area 177.8 mm
Alternate Part Number J50-265-2595
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.