Learn More
Atago™ PR-301α Model Digital Refractometer
Click to view available options
Unit Size:
Each
Description
- Digital refractometer
- Improved measurement accuracy
- Compact and battery operated
- Display the measured values of samples quickly
Specifications
Specifications
| Type | Digital Refractometer |
| Scale Range | 45 to 90 Brix |
| Model | PR-301α |
| Accuracy | ±0.1% Brix at 10 to 30°C |
| Resolution | 0.1% Brix |
| Scale Type | Brix |
| For Use With (Application) | The measurements of jellies, raw liquid sugar |
| Includes | Standard liquid LG, Round sheet |
| IP Rating | IP64 |
| Weight (Metric) | 300 g |
| Show More |