Learn More
RETSCH PP 25 Pellet Press
Prepare solid samples for XRF analysis with this pellet press. Retsch™ PP 25 Pellet Press is a compact benchtop unit with particularly simple and safe operation. Produces high quality pellets characterized by their high degree of stability.
Brand: RETSCH 20.750.0006
This item is not returnable.
View return policy
Description
- Produces high quality, stable pellets
- Easy and safe operation
- Several pressing tools available
- Can be evacuated
- Compact benchtop unit
Specifications
Pellet Press | |
Polypropylene | |
0.8 kg |
RET077 Pellet Press PP25
File size: 149.5 kB
Product Content Correction
Product Title
RETSCH PP 25 Pellet Press > Pellet Press