missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Poxygrid Steel Test Tube Racks
Shop All SP Scienceware Products
Click to view available options
Color:
Blue
Green
Orange
Yellow
Array:
4 x 10
4 x 12
6 x 12
8 x 10
9 x 12
Dimensions (L x W x H):
20.3 x 9.2 x 6.4 cm
20.5 x 16.4 x 6.4 cm
24.2 x 13 x 6.4 cm
24.2 x 9.2 x 6.4 cm
24.3 x 11 x 8.3 cm
24.3 x 20.3 x 8.3 cm
26.3 x 11.6 x 8.3 cm
29.3 x 12.8 x 8.3 cm
29.3 x 24.2 x 8.3 cm
For Use With (Equipment):
10 - 13mm Tubes
13 - 16mm Tubes
16 - 20mm Tubes
20 - 25mm Tubes
Unit Size:
Each
Description
Chemical and corrosion resistant steel test tube racks in different colors for easy identification.
- Chemical and corrosion-resistant steel wire racks are coated with heat-cured epoxy
- Orange racks can be used for biohazard warning, yellow for radioactive warning
- Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
- Will withstand a temperature range of -223°C (-369°F) to 121°C (250°F)
- All racks have square holes, three decks, a slotted grid bottom and reinforced edges
Specifications
Specifications
| Product Type | Test Tube Rack |
| Array | 8 x 10 |
| Autoclavable | Autoclavable |
| Color | Yellow |
| Holds | Tubes (20 to 25 mm) |
| Material | Epoxy-coated Steel |
| Positions | 80 |
| Shape | Rectangular |
| Sterility | Non-sterile |
| For Use With (Equipment) | 20 - 25mm Tubes |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction