Learn More
Brady™ PowerMark™ Polyester Labeling Tape
Click to view available options
Color:
Orange
Transparent
White
Yellow
Width (Metric):
101.6 mm
152.4 mm
177.8 mm
254 mm
Unit Size:
Each
Description
- For labelling stainless steel and smooth surfaces.
- Applies best to very smooth surfaces and ideal for mounting to rigid panels.
- B-569 adheres to smooth surfaces with the same durability as B-595.
Specifications
Specifications
| Certifications/Compliance | Compliant with Directive 2011/65/EU and 2015/863 |
| Description | BBP85 Tape - B-569 178 mm Yellow |
| Adhesive Type | Permanent Acrylic, Pressure-sensitive |
| Coating | None |
| Color | Yellow |
| Finish | Gloss |
| Format | Continuous |
| Material | Polyester |
| Printable Sides | Single-side Printable |
| Print Technology | Thermal Transfer |
| Show More |