Learn More
Invitrogen™ PowerEase™ Touch 120W Power Supply and XCell SureLock™ Mini-Cell Welcome Pack
The PowerEase Touch 120W Power Supply and XCell SureLock Mini-Cell Welcome Pack contains: PowerEase Touch 120W Power Supply and XCell SureLock Mini-Cell
Brand:  Invitrogen™ PSC120S
1013.20 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
Product Code. 17146961
  • € 1192.00 / Each
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Description
The PowerEase Touch 120W Power Supply and XCell SureLock Mini-Cell Welcome Pack contains:
  • PowerEase Touch 120W Power Supply
  • XCell SureLock Mini-Cell
About the PowerEase Touch 120W Power Supply
The Invitrogen PowerEase Touch 120W Power Supply is a medium-throughput power supply with a 4.3-inch backlit LCD touchscreen. The touchscreen and user interface design make setting up custom protocols or selecting one of the several pre-programmed gel electrophoresis and transfer methods a breeze. This power supply is ideal for DNA/RNA electrophoresis, SDS-PAGE, and native PAGE. The PowerEase Touch 120W Power Supply offers four sets of output jacks that can be used simultaneously and three modes: constant voltage, constant current, and constant power for flexibility of use and efficiency.
About the XCell SureLock Mini-Cell
The Invitrogen XCell SureLock Mini-Cell is a vertical mini-protein gel electrophoresis system compatible with all Invitrogen mini precast gels and Surecast handcast gels. The Mini-Cell is simple, sturdy, and easy to use. Rather than using clamps, the system incorporates the use of a gel tension wedge to make setup quick and convenient.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PowerEase
1 pack
Room temperature