Learn More
Robin™ Power Mixers
Product Code. 11969607
Click to view available options
Speed Range:
12 t/min.
2 to 25 t/min.
2 to 40 t/min.
Unit Size:
Each
Product Code. 11969607
Brand: Robin™ PM032
Only null left
Specifications
Speed Range 2 to 40 t/min.
Voltage 230 V
Holds 2 L
Product Type Power Mixer
Frequency 50 Hz
Depth (Metric) 34 cm
Height (Metric) 48 cm
Length (Metric) 38 cm
Weight (Metric) 6 kg