Learn More
Edmund Bühler GmbH™ Power Control LR 1
Provides stepless control of the current consumption between 0 to 100%. Edmund Bühler GmbH™ Power Control LR 1 is for use with IRB 1 Infrared Heater.
Brand:  Edmund Bühler GmbH™ 6070 000
Product Code. 10160761
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • For use with IRB 1 Infrared Heater
  • Provides stepless control of current consumption from 0 to 100%.
Specifications
Specifications
IRB 1 Infrared Heater
Power Control LR 1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers