Learn More
Pour Point Standard, Paragon Scientific™
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
Description
- Certified according to ISO 17025 / ISO 17034 under UKAS accreditation by means of a method specific inter-laboratory study.
- Fully traceable to international standards.
- Supplied with MSDS.
- Tamper evident security packaging.
- 12 month shelf life.
- Manufactured in the United Kingdom
Specifications
Specifications
| Certifications/Compliance | ISO 17025, UKAS ISO 17034 |
| Quantity | 250 mL |
Nominal values quoted