Learn More
Memmert™ Potential-Free Contact (24V/2A) W/Socket To Namur Ne 28 For Combined Error Message, For Ich750
Shop All Memmert Products
Click to view available options
Voltage:
24 V
Unit Size:
Each
Description
Includes
Socket 3-Pin
Specifications
Specifications
| For Use With (Equipment) | Models VO, VOcool, HCP, TTC, CTC, U, UF TS, UNpa, S, I, ICP, IPP, IPS, HPP, ICH, 108 |
| Type | Potential-free Contact |
| Voltage | 24 V |
| Amperage | 2 A |