missing translation for 'onlineSavingsMsg'
Learn More

Potassium permanganate, Puriss., meets analytical spec. of BP, Ph. Eur., USP, 99 to 100.5%, Honeywell Fluka™
BIOPHARMA

Product Code. 15611280
Click to view available options
Quantity:
250 g
500 g
1 kg
2.5 kg
25 kg
6 x 1 kg
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

Puriss., meets analytical specification of BP, Ph. Eur., USP, 99-100.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-64-7
Molecular Formula KMnO4
Molecular Weight (g/mol) 158.032
MDL Number MFCD00011364
InChI Key VZJVWSHVAAUDKD-UHFFFAOYSA-N
Synonym potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
PubChem CID 516875
IUPAC Name potassium;permanganate
SMILES [O-][Mn](=O)(=O)=O.[K+]

Specifications

Quantity 1 kg
UN Number 1490
Formula Weight 158.03g/mol
Percent Purity 99 to 100.5%
Grade puriss.
Chemical Name or Material Potassium Permanganate, Meets analytical specification of Ph. Eur.BP, USP
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Potassium permanganate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Oxidising solid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 C
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : potassium permanganate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.