Learn More
Potassium Hydroxide Solution, 50%, Chem Lab™
Product Code. 11959202
Click to view available options
Quantity:
1 L
Packaging:
1L
Specifications
Color Colorless
Concentration 500g KOH/l H2O
Density 1.35g/mL
Solubility Information Soluble in water
Quantity 1 L
Odor Odourless
Packaging 1L
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • Potassium hydroxide 50 vol. % solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : potassium hydroxide