Learn More
Prestige Medical™ Portable Autoclave Classic Media
Use this autoclave where a cost effective, portable autoclave is needed. Prestige Medical™ Portable Autoclave Classic Media is fully automatic, simply press the button to start. Lightweight and portable, ideal for laboratory use. This 12 liter Classic Media autoclave sterilizes at 121°C.
Brand: Prestige Medical™ 210048
This item is not returnable.
View return policy
Description
- High quality construction
- Fully automatic electronic time and temperature control
- Steam release valve automatically opens to release excess pressure
- Automatic fault warning light and cut-out if unit boils dry or sterilisation is interrupted
- Dual interlock handles prevent the lid being opened before the pressure and temperature inside have fallen to safe levels
- Complete with instrument basket
- Models 2015 onwards available with optional printer for easy data recording
- CE marked
- Comprehensive one year warranty (UK)
- Interlock safety mechanism, eliminates the risk of super-heated vessels exploding
- Reliable, special in-built software ensures exact temperatures are reached for the correct duration
- 9L and 12L models
- Fits neatly onto the smallest of workbenches For the sterilization of media, liquids and contaminated waste