Learn More
Haldenwanger™ Porcelain Buchner Funnel
Click to view available options
Capacity (Metric):
50 mL
75 mL
135 mL
290 mL
580 mL
795 mL
Diameter (Metric) Plate:
48mm
59mm
74mm
90mm
114mm
129mm
Unit Size:
Each
Description
Designed with glazed porcelain for excellent thermal and chemical resistance. Haldenwanger™ Porcelain Buchner Funnels are an excellent choice for quick filtration.
- Vertical sides
- Available in a variety of sizes to fit many filtration needs
- Perforated fixed filter plate
Specifications
Specifications
| Capacity (Metric) | 795 mL |
| Material | Porcelain |
| Diameter (Metric) Plate | 129mm |
| Quantity | 1Pack |
| Product Type | Buchner Funnel |