Learn More
Applied Biosystems™ POP-6™ Polymer, for 3500/SeqStudio™ Flex
Product Code. 16386502 Shop All Applied Biosystems Products
Click to view available options
Quantity:
384 Samples
96 Samples
960 Samples
Unit Size:
Each
Pack of 384
Pack of 96
Pack of 960
Product Code. 16386502
Brand: Applied Biosystems™ A26071
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
The pouch contains 96 samples of the polymer
POP-4, POP-6, and POP-7 are conveniently offered in easy to use pouch packages. The POP-6 separation matrix is optimized for standard sequencing and fragment analysis.
POP-4, POP-6, and POP-7 are conveniently offered in easy to use pouch packages, POP-6 separation matrix is optimized for standard sequencing and fragment analysis. POP polymers dynamically coat the capillary wall to control electro-osmotic flow. Their definite quality and uniform consistency eliminate guesswork and help to ensure reproducibility. POP Polymers are specifically formulated to separate DNA fragments of a known size range at a desired resolution and run time.
POP-4, POP-6, and POP-7 are conveniently offered in easy to use pouch packages with multiple sample size formats. The POP-6 separation matrix is optimized for standard sequencing and fragment analysis.
Easy to use: Pre-formulated and in a single use pouch, which not only saves time but also ensures reliability and consistency
Include radio frequency identification (RFID) label: easy to track important information such as part/lot number, samples remaining, and expiry dates (displayed on software dashboard)
Robust formulation: formulated for sequencing and fragment analysis applications
Replenishable: alow capillaries to be used multiple times
Reproducible Results
POP polymers dynamically coat the capillary wall to control electro-osmotic flow. Their definite quality and uniform consistency eliminate guesswork and help to ensure reproducibility. POP Polymers are specifically formulated to separate DNA fragments of a known size range at a desired resolution and run time.
Many Choices
POP-7 polymers are very versatile and designed for a wide range of short read to long read sequencing applications as well as fragment analysis. POP-4 polymers are designed specifically for HID/Forensic applications, while POP-6 polymers are the choice for certain sequencing applications (and fragment analysis).
For partially used pouches that need to be stored, a cap (4412619) that plugs into the pouch fitment opening is recommended.
Order Info
Each
TRUSTED_SUSTAINABILITY
Specifications
Quantity 96 Samples
For Use With (Application) Standard Sequecning, Fragment Analysis (Denaturing)
Product Line POP-6