Learn More
Polyvinyl alcohol, 86-89% hydrolyzed, medium molecular weight, Thermo Scientific Chemicals
€ 22.41 - € 113.57
Chemical Identifiers
CAS | 9002-89-5 |
---|---|
Molecular Formula | (C2H4O)n |
Molecular Weight (g/mol) | 44.05 |
MDL Number | MFCD00081922 |
InChI Key | IMROMDMJAWUWLK-UHFFFAOYSA-N |
Synonym | vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval |
PubChem CID | 11199 |
IUPAC Name | ethenol |
SMILES | OC(-*)C-* |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11339607
|
Thermo Scientific Alfa Aesar
041239.14 |
25 g |
€ 22.41
25g |
In Stock
Log in to see stock. |
|||||
11329607
|
Thermo Scientific Alfa Aesar
041239.22 |
100 g |
€ 44.32
100g |
In Stock
Log in to see stock. |
|||||
11319607
|
Thermo Scientific Alfa Aesar
041239.36 |
500 g |
€ 113.57
500g |
In Stock
Log in to see stock. |
|||||
Description
Polyvinyl alcohol is used as a biological reagent to test the dispersion of zinc oxide nanoparticles. It is an effective protein resistant material and is used to stabilize gold colloids of diameter 3-8nm. It is used as an emulsion polymerization aid to make polyvinyl acetate, as a protective colloid, as a film used in the water transfer printing process, as a surfactant for the formation of polymer encapsulated nanobeads, as an embolization agent in medical procedures and as a fixative for specimen collection.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
9002-89-5 | |
44.05 | |
IMROMDMJAWUWLK-UHFFFAOYSA-N | |
11199 | |
OC(-*)C-* |
(C2H4O)n | |
MFCD00081922 | |
vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval | |
ethenol |
Specifications
9002-89-5 | |
(C2H4O)n | |
vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval | |
IMROMDMJAWUWLK-UHFFFAOYSA-N | |
ethenol | |
11199 | |
Odorless |
Powder | |
MFCD00081922 | |
Soluble in water. | |
OC(-*)C-* | |
44.05 | |
86 to 89% | |
Polyvinyl alcohol, Medium molecular weight, Hydrolyzed |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only