Learn More
mb2 Polyurethane Chair on Glides - Asynchronous Mechanism
Click to view available options
:
Height: 540mm
Description
- Chair in polyurethane
- Seat and back in polyurethane
- Asynchronous mecanism, adjustable seat and backrest
- Pneumatic cylinder low
- 5-spoke base injected in nylon and fiberglass reinforced by a galvanized steel ring.
- glides
Specifications
Specifications
| Product Type | Laboratory Seat |
| Color | Black |
| Adjustable (Height) | Yes |
| Adjustable | Height |
| Autoclavable | Not Autoclavable |
| Back Style | Standard |
| For Use With (Equipment) | For Benchtop |
| Height (Metric) | 540 mm |
| Quantity | 1 Ea. |
| Seating Height | 400 to 540 mm |
| Show More |