Description
- Knitted polyamide glove
- Black oilproof nitrile coating on palm and back
- Fully dipped
- Smooth finish
- Elastic wrist
- The knitted construction conforms perfectly to the natural shape of the hand thus providing excellent dexterity
- Open back to provide superior ventilation
- Polyamide combines good resistance to abrasion, humidity, tearing and shearing
- The nitrile coating on the palm and fingertips brings some excellent abrasion resistance quality as well as oilproof properties
- A pattern finish to enhance grip
- Black color dedicated to dirty applications and environments
Specifications
Specifications
Certifications/Compliance | EN 420-2003 Basic standards, EN 388-2016 Mechanical hazards |
Description | 13 gauge |
Coating Material | Polyamide, Nitrile |
Color | Black |
Size | 7 |
Length (Metric) | 25 cm |
Product Content Correction
Product Title
Honeywell™ POLYTRIL™ Top Gloves >