Learn More
Greiner Bio-One Polystyrene Vented Petri Dishes
Click to view available options
Sterility:
Sterile
Diameter (Metric):
35 mm
60 mm
145 mm
Height (Metric):
10 mm
15 mm
20 mm
Unit Size:
Pack of 120
Pack of 600
Pack of 740
Description
- Vented for improved gas exchange
- Manufactured from crystal polystyrene for optimal analysis by microscopy
Specifications
Specifications
| Color | Clear |
| Material | Polystyrene |
| Shape | Round |
| Sterility | Sterile |
| Type | Petri Dish |
| No. per Case | 120 |
| Vent | Yes |
| Dimensions (Dia. x H) | 145 x 20 mm |
| Height (Metric) | 20 mm |
| Diameter (Metric) | 145 mm |
For Research Use Only. Not for use in diagnostic procedures, unless otherwise indicated.