Learn More
Kartell™ Polypropylene Urine Containers
Product Code. p-8000307
Click to view available options
Sterility:
Aseptic
Non-sterile
Sterile
Closure Color:
Blue
Red
No. per Pack:
250/Pk.
500/Pk.
600/Pk.
Unit Size:
Pack of 250
Pack of 500
Pack of 600
Product Code. 10747721
Brand: Kartell™ 0564000
Edge
Manufactured from clear polypropylene for strength and chemical resistance. Kartell™ Polypropylene Urine Containers are available in sterile and non-sterile with color-coded screw caps.
  • Non sterile cups have blue screw caps; sold in multi-packs
  • Sterile cups have red screw cap and a sanitized marking area; individually wrapped
  • CE marked; IVD approved
Specifications
Closure Included Yes
Graduated Yes
Material Polypropylene
Sterility Non-sterile
Capacity (Metric) 150 mL
Certifications/Compliance CE and IVD
Closure Color Blue
Closure Material Polypropylene
Closure Type Screw Cap
Diameter (Metric) 63 mm
For Use With (Application) Urine collection
Graduations 20 to 100 mL
Height (Metric) 73 mm
Includes Polypropylene Screw Cap
No. per Pack 500/Pk.