Learn More
BRAND™ Polystyrene Slide Boxes
Shop All Brand Products
Click to view available options
Slide Capacity:
25
50
100
Unit Size:
Each
Description
- Polystyrene construction
- For use with microscope slides
- Dimensions: 76 x 26mm
Specifications
Specifications
| Slide Capacity | 25 |
| Device Holds | 25 Slides |
| Length (Metric) | 76 mm |
| Material | Polystyrene |
| Width (Metric) | 26 mm |