Learn More
Multiroir™ Polystyrene Rectangular Trays
Product Code. 11768361
Click to view available options
Height (Metric):
34 mm (Inner), 37 mm (Outer)
40 mm (Inner), 42 mm (Outer)
42 mm
Color:
Blue
Green
Red
White
Yellow
Unit Size:
Each
Product Code. 11768361
Brand: Multiroir™ 39101
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Organize your workspace with these durable trays. Multiroir™ Polystyrene Rectangular Trays are widely used in the laboratory and available in a variety of colors and styles.
  • Multi-purpose
  • Polystyrene
  • Rectangular
Specifications
Color White
Height (Metric) 34 mm (Inner), 37 mm (Outer)
Length (Metric) 175 mm (Inner), 205 mm (Outer)
Material Polystyrene
Type Tray
Width (Metric) 100 mm (Inner), 130 mm (Outer)
No. per Pack 1 Pack
Shape Rectangular