Learn More
Multiroir™ Polystyrene Rectangular Trays
Product Code. 11738371
Click to view available options
Height (Metric):
34 mm (Inner), 37 mm (Outer)
40 mm (Inner), 42 mm (Outer)
42 mm
Color:
Blue
Green
Red
White
Yellow
Unit Size:
Each
Product Code. 11738371
Brand: Multiroir™ 39203
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Organize your workspace with these durable trays. Multiroir™ Polystyrene Rectangular Trays are widely used in the laboratory and available in a variety of colors and styles.
  • Multi-purpose
  • Polystyrene
  • Rectangular
Specifications
Color Yellow
Height (Metric) 40 mm (Inner), 42 mm (Outer)
Length (Metric) 210 mm (Inner), 260 mm (Outer)
Material Polystyrene
Type Tray
Width (Metric) 150 mm (Inner), 196 mm (Outer)
No. per Pack 1 Pack
Shape Rectangular