Learn More
Multiroir™ Polystyrene Rectangular Trays
Product Code. 11718371
Click to view available options
Height (Metric):
34 mm (Inner), 37 mm (Outer)
40 mm (Inner), 42 mm (Outer)
42 mm
Color:
Blue
Green
Red
White
Yellow
Unit Size:
Each
Product Code. 11718371
Brand: Multiroir™ 39201
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Organize your workspace with these durable trays. Multiroir™ Polystyrene Rectangular Trays are widely used in the laboratory and available in a variety of colors and styles.
  • Multi-purpose
  • Polystyrene
  • Rectangular
Specifications
Color White
Height (Metric) 42 mm
Length (Metric) 260 mm
Material Polystyrene
Type Tray
Width (Metric) 196 mm
No. per Pack 1 Pack