missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Polystyrene Petri Dishes

Product Code. 10161573
Click to view available options
Diameter (Metric):
90 mm
120 mm
Shape:
Round
Square
Surface Treatment:
Aseptic
Beta-Ray Sterilization
Includes:
4 Sections
Triple Vents
Unit Size:
Pack of 180
Pack of 500
This item is not returnable. View return policy

Product Code. 10161573

Brand: Kartell™ 0036600

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Constructed from crystal clear polystyrene for optical clarity. Kartell™ Polystyrene Petri Dishes are suited for routine lab work and bacteriologial research.

  • Disposable
  • Clear Petri dish constructed of PS (Polystyrene)
  • Designed for routine lab work
  • Can be used in bacteriological research
  • CE Marked, IVD Approved

Specifications

Certifications/Compliance CE/IVD
Color Clear
Disposable Single-use
Material Polystyrene
Shape Round
Sterility Sterile
Surface Treatment Beta-Ray Sterilization
For Use With (Application) For routine lab work, bacteriological research
Includes Triple Vents
No. per Pack 500
Vent Yes
Diameter (Metric) 90 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.