Learn More
Greiner Bio-One Polystyrene Petri Dishes
Product Code. 11759452
Click to view available options
Dimensions (Dia. x H):
35 x 10 mm
60 x 15 mm
94 x 15 mm
94 x 16 mm
100 x 20 mm
120 x 17 mm
145 x 20 mm
Sterility:
Non-sterile
Sterile
Vent:
No
Yes
Quantity:
120 Pack
240 Pk.
360 Pack
480 Pack
560 Pack
600 Pack
740 Pack
Unit Size:
Pack of 120
Pack of 240
Pack of 360
Pack of 480
Pack of 560
Pack of 600
Pack of 740
Product Code. 11759452
Brand: Greiner Bio-One 633179
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured from highly transparent polystyrene for its heat resistant properties to enable use with hot agar. Greiner BioOne™ Polystyrene Petri Dishes are available in a wide variety of formats and sizes to accommodate multiple research needs.
  • Heat resistant up to 60°C
  • Available in vented and non-vented
  • Choose from standard round design, square-profile dish, special models and contact dishes/OneWell Plate™ models
  • Easy stacking
  • Crystal polystyrene for optimal analysis by microscopy
TRUSTED_SUSTAINABILITY
Specifications
Closure Type Stackable
Disposable Single-use
Material Polystyrene
Shape Round
Sterility Sterile
Quantity 480 Pack
Vent Yes
Dimensions (Dia. x H) 94 x 15 mm
Height (Metric) 15 mm
Diameter (Metric) 94 mm