Learn More
Fisherbrand™ Polystyrene Macro Cuvettes for UV and Visible Wavelengths
Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
4 mL
Unit Size:
Pack of 1000
Specifications
Specifications
| Type | Macro Cuvette |
| Disposable | Reusable |
| Capacity (Metric) | 4 mL |
| Material | Plastic |
| Quantity | 100/Pk. |
| Wavelength Range | 220 to 900 nm |
| Height (Metric) | 45 mm |
| Width (Metric) | 12.5 mm |